Hypersil GOLD™ PREP C18 HPLC Columns, Thermo Scientific™
Supplier: Thermo Scientific
Certificates
About this item
Thermo Scientific™ Hypersil GOLD™ C18 preparative HPLC columns are used to maintain exceptional peak shape and resolution for scaling up purification needs.
- Proprietary functionalization and end-capping
- C18 (USP L1) selectivity
- Robust performance at high pH
- Excellent column stability at low pH
- Highly symmetrical peak shapes
- Improved speed of analysis, efficiency, and sensitivity
These columns come in a variety of dimensions and an option for XtendedLife hardware which improves column lifetime by maintaining a uniform packed bed. These endcapped, ultrapure, silica-based columns deliver significant reduction in peak tailing using generic gradients with C18 selectivity. Hypersil GOLD columns give you confidence when scaling up from your analytical method.
Specifications
- Catalog No:
- 76332-200
- 76332-194
- 76332-196
- 76332-190
- 76332-192
- 76332-186
- 76333-940
- 76332-188
- 76332-154
- 76332-152
- 76332-158
- 76332-156
- 76332-146
- 76332-150
- 76332-202
- 76332-148
- 76332-162
- 76332-160
- 76332-164
- 76332-138
- 76332-176
- 76332-178
- 76332-142
- 76332-172
- 76332-370
- 76332-140
- 76332-174
- 76332-168
- 76332-170
- 76332-166
- 76332-184
- 76332-144
- 76332-180
- 76332-182
- Carbon load:
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- 10%
- pH range:
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- 2 - 9
- Surface area:
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g
- 220 m²/g