VWR® UV/Visible Spectrophotometer, P9
# de catalogue 76644-700
Fournisseur: VWR International
Spécifications
- Absorbance range:–0,3 - +3,0 A
- Light source:Tungsten/deuterium
- Description:UV/Visible spectrophotometer
- Model:P9
- W×D×H:580×420×235 mm
- Wavelength accuracy:±0.3 nm
- Photometric range:0 - 200% T
- Scan speed:6000 nm/min
- Spectral bandwidth:0.5/1/2/4/5 nm
- Wavelength repeatability:≤0.2 nm
- Weight:18 kg
- Spectral Range:190 - 1100 nm
- Cat. no.:76644-700
Spécifications
A propos de cet article
The P9 UV/Visible double-beam spectrophotometer is an easy to use instrument with advanced performance.
- Highly accurate and reliable
- Simple, elegant design with aluminum base
- Easy to change lamp
- User friendly interface
- 64 GB internal storage capabilites
- USB-A + B, Ethernet, VGA, HDMI connectivity
Provides several basic functions, such as photometry test, multi wavelength scanning, kinetics, quantitation test, spectrum scanning and biological methods.
Ordering information: More accessories are available and also many parts from the first generation can be used as well. For more information please contact the Avantor quotation team or your local Avantor sales representative.
Delivery: The instrument is equipped with a manual 4-position cell changer and is shipped with software, sample cuvettes (glass/quartz), power cords (EU/UK/CH), USB cable and dust cover.
Customers Who Bought This Also Bought