SealPlate® RollMate™, Pre-Secored Sheets on a Roll with Dispenser, Non-Sterile, Excel Scientific
Catalog # 76293-222
Supplier: VWR International
Specifications
- Optical property:Transparent
- Sterility:Non sterile
- Description:Sealing film, polyester, 500 sheets/roll
- Thickness:77,5 µm
- Cat. no.:76293-222
Specifications
About this item
The SealMate SealPlate Starter Kit features a re-usable dispenser and two sterile rolls of perforated film.
The dispenser facilitates the quick and easy dispensing of film from a roll allowing microplates to be sealed three times faster than using sheeted films, while protecting unused seals from contamination. Cost-effective replacement rolls are sold separately. Ideal for use under the hood; the SealMate dispenser minimizes film contamination.
Customers Who Bought This Also Bought