BioClean™ Nitrile Clean Non Sterile RABS/Isolator Gloves, Ansell
Fournisseur: Ansell Healthcare
Certificats
A propos de cet article
Non sterile RABS/Isolator gloves.
- Tested against ASTM D6978-05 for handling chemo drug
- 100% water leak tested
- Ultra-clean surface ensures product protection
- Autoclave compatible
- ISO 4 Cleanroom laundered and packaged
- Resistant to VHP, IPA, and disinfectants
BioClean™ RABS and Isolator Gloves are manufactured from nitrile with incredibly low levels of particles and excellent ESD properties. Designed for use in injectable manufacturing and pharmaceutical production areas, the CGL series of gloves are cleanroom processed and packed, and are available in a range of port sizes.
Certifications: ASTM D6978, CE 0598, EC 1935/2004, EN 388:2016, EN 420:2003 + A1:2009, EN 421:2010, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 455 Part 2, Category III.
Packaging: One piece per sealed inner PE bag; one inner bag per sealed second inner PE bag; one second inner bag per sealed outer PE bag; 20 outer bags per lined inner white Correx polyethylene box (20 pieces).
Spécifications
- Catalog No:
- 76316-253
- 76316-257
- 76316-255
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Material:
- Nitrile
- Nitrile
- Nitrile
- Finish:
- Non-clean, fully dipped
- Non-clean, fully dipped
- Non-clean, fully dipped
- Cuff style:
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 20 mil
- 20 mil
- 20 mil
- Palm thickness:
- 20 mil
- 20 mil
- 20 mil