A propos de cet article
The Rapid Response™ Xylazine Test Strip (Liquid / Powder) detects the presence xylazine in liquid or powder samples at a cut-off concentration of 1000 ng/mL or higher.
- Sample: Liquid/Powder
- Quantity: 100 Tests/Kit
- Detects xylazine at concentrations of 1000 ng/mL and above
- Results as soon as 60 seconds
- Results interpretation guide printed on each pouch
- Easy to interpret results with built-in procedural controls
- Individually packaged strips for easy distribution
Warning: This test does not evaluate drug safety or purity.
The Rapid Response™ Xylazine Test Strip (Liquid / Powder) is a rapid visual immunoassay for the qualitative, presumptive detection of xylazine in liquid or powder substances at a cut-off of 1000 ng/ml or higher.
This kit includes individually packaged test strips with result interpretation guide printed on the back of each pouch, a result interpretation card to aid in reading the results, and detailed instructions.
To perform the test, remove the test strip from the pouch and hold the strip at the end where the product name is printed. Dip the test strip into liquid or powder sample (diluted in water) for 10-15 seconds. Remove the strip from the sample and place it on a non-absorbent flat surface. A negative result can be interpreted as soon as both the test (T) and control (C) lines appear. A positive result can be interpreted once the control line has appeared and the background has cleared to white, typically by 60 seconds. Do not read results after 10 minutes. Please read the Product Insert supplied with the kit before use.
The test device is not intended to determine the purity, composition, or if the substance being examined is safe to use. A positive or negative test result is not an indication that the substance being examined is safe to use. Many factors come into play when examining the samples, including but not limited to a mixture of multiple substances, solubility, and pH of the sample. BTNX does not encourage the use, supply, or production of illegal drugs or controlled substances in any way. The device is intended for harm reduction purposes. Follow the advice of local harm reduction or public health agencies. Not for testing Cocaine.
There is a possibility that technical or procedural errors as well as other substances and factors may interfere with the Rapid Response™ Xylazine Test Strip (Liquid / Powder) and cause false results. A positive result indicates the presence of xylazine only and does not indicate quantity. A negative result does not at any time rule out the presence of xylazine, as they may be present below the minimum detection level of the test.