Saf-T-Trainer Cartridges
Fournisseur: PRECISION GUN SPECIALTIES
Certificats
A propos de cet article
Bring Cartridges into the classroom safely.
- Safety orange color easily identifies as inert
- Available in multiple calibers
- Use for basic comparison, or crime scene staging
Inert training cartridges are developed for military and law enforcement use, when training or instructional activities require use of a dummy cartridge. Manufactured from a durable synthetic material and colored 'safety orange' for instant identification. An affordable replacement for traditional metallic dummy cartridges that are drilled.
Customers Who Bought This Also Bought