GM-171 Stereo Microscopes, Motic
Fournisseur: VWR International
Certificats
A propos de cet article
The GM-171 is a sophisticated microscope ready for the demands of jewelers and gem dealers on the examination of gems and precious stones. The magnification is handled by the optical body of Motic’s most recently introduced stereo microscope SMZ-171, available in binocular or trinocular version.
- Ergonomic setup with clever functionality
- Microscope stand is tiltable from 0° (upright) to 45° to suit individual seating positions and rotatable by 360° to share visual insights
- Interpupillary distance can be set from 52 - 75 mm
- Works perfectly fine with the new 10X widefield eyepieces and the zoom body of 0.75X up to 5X magnification
The GM171 is an advanced solution for jewelry examination, delivering reliable results and adaptable to a wide range of individual requirements.
Ordering information: Contact your Avantor representative to learn more about these and other motic solutions.
Customers Who Bought This Also Bought