Eisco® Slotted Masses, Zinc Casted
Fournisseur: Wards
Certificats
A propos de cet article
Individual Slotted Masses for placement on hangers are made of powder-coated, die-cast zinc and stamped with mass value for easy identification.
- Masses die-cast in zinc and powder-coated to ensure durability and long life
- Slotted for stacking on mass hangers (available separately)
- Each piece is stamped with mass value for easy identification
- Accuracy of ±2% make them an economical alternative to brass masses
- Ideal as additional or spare part for slotted mass sets
Individual Slotted Masses are made of cast zinc and powder-coated to ensure durability and long life. Each piece is slotted for stacking on mass hangers (not included) and is stamped with mass value for easy identification. Great as replacements for missing pieces or as spares.
Customers Who Bought This Also Bought