Difco™ Phytone™ Supplement, Ultra-Filtered (UF), Gibco™ (a part of Thermo Fisher Scientific)
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Difco Phytone, ultra-filtered (UF), is an animal origin-free enzymatic digest of soy. Due to the naturally high concentrations of vitamins and carbohydrates in soy, Difco Phytone UF is a nutritious and excellent carbohydrate source.
- Ultra-filtered
- Animal origin-free nutritional supplement
- Excellent carbohydrate source due to the naturally high concentrations of vitamins and carbohydrates in soy
- Desirable for use with mammalian cell culture applications where reduced endotoxin levels are important
- Recommended for use in media for the cultivation of a wide variety of organisms, including fungi
This product is an ultra-filtered version of Phytone peptone, making it desirable for use with mammalian cell culture applications where reduced endotoxin levels are important.
Customers Who Bought This Also Bought