Bacto™ Proteose Peptone No. 3, Gibco™ (a part of Thermo Fisher Scientific)
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Bacto Proteose Peptone No. 3 is a member of the Bacto Proteose Peptones, which are enzymatic digests of protein.
- Bacto Proteose Peptone is used in preparing microbiological culture media and in producing bacterial toxins
- Bacto Proteose Peptone No. 2 is used in preparing microbiological culture media
- Bacto Proteose Peptone No. 3 is used in preparing microbiological and mammalian cell culture media
- Bacto Proteose Peptone No. 4 is a spray-dried version of Bacto Proteose Peptone
Bacto Proteose Peptone No. 4 offers the same beneficial nutrients as Bacto Proteose Peptone for growth promotion and toxin production in a wide range of fastidious microorganisms.
Certifications: LUO, MUO.
Customers Who Bought This Also Bought