VWR® Cleanroom Gloves, Nitrile, Solid White, Tacky, Class 10 / ISO 4
Supplier: VWR International
Certificates
About this item
VWR® cleanroom nitrile gloves are manufactured from a synthetic copolymer of acrylonitrile, butadiene and methylacrylic acid, that provides reliable, durable and comfortable hand protection during demanding cleanroom applications.
- Beaded cuff
- Nitrile
- Silicone-free
- Low particle count
- ESD performance: 10⁹ ohms (CAFÉ), 10¹¹ ohms (Surface), 10⁹ ohms (Volume)
Certifications: N ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016; CAT III; AQL 1.5
Packaging: 100 gloves per double bag, 10 bags per case.
Specifications
- Catalog No:
- 76533-210
- 76533-208
- 76533-206
- 76533-216
- 76533-214
- 76533-212
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Finish:
- Tacky
- Tacky
- Tacky
- Tacky
- Tacky
- Tacky
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Cuff thickness:
- 0.09 - 0.12 mm
- 0.09 - 0.12 mm
- 0.09 - 0.12 mm
- 0.09 - 0.12 mm
- 0.09 - 0.12 mm
- 0.09 - 0.12 mm
- Finger thickness:
- 0.13 - 0.18 mm
- 0.13 - 0.18 mm
- 0.13 - 0.18 mm
- 0.13 - 0.18 mm
- 0.13 - 0.18 mm
- 0.13 - 0.18 mm
- Palm thickness:
- 0.11 - 0.15 mm
- 0.11 - 0.15 mm
- 0.11 - 0.15 mm
- 0.11 - 0.15 mm
- 0.11 - 0.15 mm
- 0.11 - 0.15 mm
- Tensile strength:
- ≥18 MPa
- ≥18 MPa
- ≥18 MPa
- ≥18 MPa
- ≥18 MPa
- ≥18 MPa
- Ultimate elongation:
- ≥500%
- ≥500%
- ≥500%
- ≥500%
- ≥500%
- ≥500%
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Anti-static:
- +
- +
- +
- +
- +
- +