About this item
This urine collection containers with needle will keep a clean and secure sample transfer directly into urine tubes.
- Closed system allows sample material to be transported, analyzed and stored without risk of contamination or leakage
- Suitable for application of evacuated blood collection tubes with an external diameter less than 18 mm
- Leak-resistant container features tight seal to help avoid spillage
The container features molded patient ID label and molded graduations in both ounces and milliliters.