Value Plastics® Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 1000/PK
# de catalogue MFLX45509-04BLK
Fournisseur: VALUE PLASTICS TE
Spécifications
- Connection type:Threaded Nut
- Description:Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 1000/PK
- Warranty:3 months
- Cat. no.:MFLX45509-04BLK
- Fitting Material:Black Nylon
- Flow Pattern:Straight
Spécifications
A propos de cet article
Best for use with panel mount fittings.
- Connection Type: panel mount lock nut, ¹/₄-28 UNF
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Optimize your application with these luer fitting accessories.
- Connection type:Threaded Nut
- Description:Fitting, Black Nylon, Panel Mount Lock Nut, 1/4-28 UNF; 1000/PK
- Warranty:3 months
- Cat. no.:MFLX45509-04BLK
- Fitting Material:Black Nylon
- Flow Pattern:Straight
Customers Who Bought This Also Bought