A propos de cet article
blueGel™ is safe, fast, compact, durable, and easy to use.
- Fully stackable for easy storage in included pouch
- Run up to 26 samples per 20 ml gel
- Integrated power supply
- Direct photo documentation with included Fold-a-View™ imaging hood
- Real-time band visualization while running your gel
- Safe nucleic acid staining with SeeGreen™, GelGreen® or SYBR-family dyes
- Error-proof gel pouring with included leak-free casting system
By integrating gel electrophoresis and blue light transillumination into an all-in-one device, you can see results instantly while still running your gel. There is no need for a separate imaging system, time-consuming staining and destaining, or harmful ethidium bromide and UV light.
The GlowRack™ tube adapter is an alternative to the P51™ Molecular Fluorescence Viewer when performing BioBits® and P51™ Molecular Glow Labs™. The GlowRack™ tube adapter fits directly into your blueGel™ electrophoresis system and serves as both a tube holder and blue light filter for performing BioBits® and P51™ Molecular Glow Labs™. Pipette directly into tubes while visualizing fluorescence using your existing blueGel™ electrophoresis system’s blue light illuminator.