Essentials LiquiThief Liquid Sampler, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Reduce cross contamination while sampling low viscosity liquids and oils.
- Manufactured in Class 100,000 cleanroom and individually packaged
- Constructed of chemically inert high-density polyethylene (HDPE)
The LiquiThief liquid samplers are designed to easily take liquid samples with the added convenience of not having to clean the sampler prior to use. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness and they are individually bagged. Choose from sterile or non sterile samplers. Sterile samplers are sterilized via gamma irradiation (25k Gy minimum).
Customers Who Bought This Also Bought