Essentials Scrapers, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Easily remove stuck on residue.
- Constructed from durable high-density polyethylene (HDPE)
- Cleanroom class 100,000 produced and individually packaged
- Disposable scrapers are ideal for food and pharmaceutical applications
These essentials scrapers are ideal for food, creams, and general scaping. Disposable scapers are molded and packaged in class 100,000 cleanroom and individually bagged to avoid cross contamination.
Customers Who Bought This Also Bought