Essentials Ladles, Sterile, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Reduce cross contamination while sampling liquids.
- Manufactured in Class 100,000 cleanroom and individually packaged
- Designed to go through traditional 2" bungs
These ladles are designed to easily take liquid samples with the added convenience of not having to clean the sampler prior to use. The ladles are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness and they are individually bagged. Samplers are sterilized via gamma irradiation (25 to 45 kGy). Choose from polypropylene or high-density polyethylene (HDPE) construction for a wide range of applications.
Customers Who Bought This Also Bought