Essentials PowderThief, Sterile HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Quickly and easily take a sample at a specified point within the vessel.
- Manufactured in Class 100,000 cleanroom and individually packaged
- Constructed of chemically inert high-density polyethylene (HDPE)
- Sterile
These PowderThief powder samplers are designed to take representative samples from known locations within the bulk powder. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness and they are individually bagged. Samplers are also sterilized via gamma irradiation (minimum dose 25 kGy).
Customers Who Bought This Also Bought