Essentials Slot Samplers, Sterile, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Reduce cross contamination while sampling various powders, granules, and grains.
- Suitable for sampling from all layers of the container, providing an accurate representation of contents
- Constructed of chemically inert high-density polyethylene (HDPE)
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. The samplers are manufactured, assembled, and packed in a Class 100,000 cleanroom to ensure cleanliness. Samplers are also sterilized via gamma irradiation (25 to 45 kGy) and individually bagged to reduce contamination.
Customers Who Bought This Also Bought