Essentials Cleaning Brushes, Antylia Scientific
Supplier: Argos Technologies
Certificates
About this item
Long handle brushes allow for a thorough cleaning.
- Brush heads conveniently screw on to the handle
- Constructed with a sturdy aluminum handle and nylon bristles
These cleaning brushes are ideal for cleaning both stainless steel, plastic, and glass samplers. Brushes heads are made with nylon Bristles in a twisted wire frame and are 100 mm long. The durable aluminum handles come in 500 mm lengths and screw together for a total handle length of 1000 mm.
Two brush head sizes are available. The 35 mm diameter brush is suitable for cleaning a vessel with a 15 to 30 mm diameter. The 50 mm diameter brush is suitable for cleaning a vessel with a 30 to 45 mm diameter.
Delivery: Brush head and two 500 mm handles.
Frequently Bought Together