Essentials Slot Samplers, Metal, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Reliably sample various powders, granules, and grains.
- Suitable for sampling from all layers of the container, providing an accurate representation of contents
- Choose from stainless steel or aluminum construction
- Nonsterile
These slot samplers provide highly representative sampling of powders, granules, and grains. They are suitable for sampling from all layers of the container, providing an accurate representation of the contents. These samplers come in different lengths, with different numbers of openings, and are made from different material options for all different sampling methods.
The 316L stainless steel construction is durable and ideal when chemical resistance is required. The lightweight aluminum construction means even the long samplers are easy to operate and manage.
Customers Who Bought This Also Bought