Essentials Powered Ice Samplers, 316L Stainless Steel, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Ideal sampling tool for wax and frozen products.
- Designed to fit most standard electric drills
- High-quality 316L stainless steel construction
Quickly cut through hard, frozen samples by attaching the ice sampler to an electric hand drill. Once powered on, the sharp-edged sampler rotates and cuts the material into the sampler. Crevice-free, highly polished surface sampler is easy to clean.
Certifications: Supplier certificate of conformity is included with product shipment.
Customers Who Bought This Also Bought