Essentials Long Handled Spoons, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Long handle minimizes potential contact with sample material.
- Easily scoop from a distance without losing spoon inside vessel
- Constructed of chemically inert high-density polyethylene (HDPE)
- Molded and packaged in Class 100,000 cleanroom and individually bagged
The long handle spoons are ideal for acquiring samples from bottles, jars, and other confined areas while helping to avoid personal contact with sample materials. All spoons are individually bagged and can be used straight from the bag without the need for costly cleaning. Choose from sterile or non-sterile spoons. Sterile spoons are gamma irradiated with a minimum dose of 25 kGy.
Customers Who Bought This Also Bought