Essentials Double-Ended Spoons, 304 Stainless Steel, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Take different sample sizes from the same spoon.
- 304 stainless steel (SS) construction for excellent chemical compatibility
- BSE/TSE free
- Non sterile
Simply flip around the spoon to take different sample sizes. Spoons are constructed of 304 SS with a polished smooth finish—ideal for powders and liquid.
Certifications: Supplier Certificate of Conformity is included with product shipment.
Customers Who Bought This Also Bought