Essentials Disposable Volumetric Spoons, Sterile, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Manufactured and packed in Class 100,000 cleanroom to ensure cleanliness and reduce cross contamination.
- Volumetric spoons accurately measure set volumes
- Made from high-quality HDPE which conforms to FDA, EC, and EU regulations
- SAL 10⁻⁶ compliant
- BSE/TSE free
Use these sterile, disposable volumetric spoons to measure and sample powders, granules, liquids, gels, and suspensions. Spoons are gamma irradiated and individually packaged so they can be used straight out of the bag without cleaning. Long handles ensure hands are away from the materials being measured.
Certifications: Supplier Certificate of Conformity is included with product shipment.
Customers Who Bought This Also Bought