Essentials Powder Lances, HDPE, Antylia Scientific
Fournisseur: Argos Technologies
Certificats
A propos de cet article
Quickly and easily take a cross sectional sample within the vessel.
- Manufactured in Class 100,000 cleanroom and individually packaged
- Ergonomically designed handle makes it easy to use, even when doing a large amount of sampling
- Constructed of chemically inert high-density polyethylene (HDPE)
These powder lance samplers are designed to take a cross sectional sample at an exact position within the vessel. The samplers are manufactured, assembled, and packed in a Class 100,000 Cleanroom to ensure cleanliness and individually bagged. Choose from sterile or non-sterile samplers. Sterilized powder lances are sterilized via gamma irradiation (25 - 45 kGy).
Customers Who Bought This Also Bought