Specifications
- Description:COSA™ Foam 40
- Size:1 gal.
- Cat. no.:76637-528
Specifications
About this item
COSA™ Foam 40 is a liquid, neutral pH product for manual cleaning, or for use as a foam additive in cleaning solutions with Cosa™ CIP products for the pharmaceutical and personal care industries.
- Suitable for manual and foam cleaning
- Clear solution at all temperatures
- Low toxicity high LD50-value and PDE value
- cGMP manufactured - Global formulation
Safe for manual use, COSA™ Foam 40 acts as a mild and odorless detergent.