Cleanroom Papers, Columbia Cleanroom
Fournisseur: Columbia Printing & Graphics
Certificats
A propos de cet article
White, 8.5×11" cleanroom paper with colored bar or edge to distinguish between departments, operations or projects.
- Class 10
- No chance for ink bleeds
- Latex-free
- Cellulose and low-density polyethylene co-polomers
Packaging: Double-bagged in packs of 250 sheets. 10 packs of 250 sheets/case.
Customers Who Bought This Also Bought