ABS® Undercounter and Countertop Refrigerators, Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
These space-saving refrigerators offer premium protection for vaccine storage.
- Microprocessor controller with LED display
- Integrated audible and visual alarms
- Hermetic, high performance compressor
- White powder coated steel construction
- Natural hydrocarbon refrigerants
- NSF/ANSI 456 Vaccine Storage Certification
- ENERGY STAR certified*
These premier undercounter and countertop refrigerators are designed in accordance with the NSF/ANSI 456 Standard for Vaccine Storage. Units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery. The refrigerators utilize microprocessor controllers and feature temperature alarms, remote alarm contacts, and probe access ports with included probes. Units utilize HFC-free refrigerant for environmental health and energy efficiency.
: Compliant with the thermal performance requirements as defined in the NSF/ANSI 456 Standard for Vaccine Storage for all testing protocols. UL, C-UL, ETL, C-ETL listed (either single or dual agency listings) and certified to UL471 standard, hydrocarbon refrigerant safety.
*Select models ENERGY STAR certified: 76637-324, 76637-326, 76637-296, and 76637-298
*Select models ENERGY STAR certified: 76637-324, 76637-326, 76637-296, and 76637-298
- Catalog No:
- 76637-322
- 76637-300
- 76637-320
- 76637-328
- 76637-324
- 76637-298
- 76637-296
- 76637-326
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76637-322
- 76637-300
- 76637-320
- 76637-328
- 76637-324
- 76637-298
- 76637-296
- 76637-326
- Temperature:
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- Four shelves (three adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Three shelves (two adjustable/one fixed)
- Three shelves (two adjustable/one fixed)
- Three shelves (two adjustable/one fixed)
- Three shelves (two adjustable/one fixed)
- Door type:
- 1 solid door, left hinged
- 1 glass door
- 1 solid door
- 1 solid door
- 1 solid door
- 1 glass door, left hinged
- 1 glass door
- 1 solid door, left hinged
- Weight:
- 40 kg (88 lbs.)
- 45 kg (100 lbs.)
- 40 kg (88 lbs.)
- 40 kg (88 lbs.)
- 43 kg (94 lbs.)
- 43 kg (94 lbs.)
- 43 kg (94 lbs.)
- 43 kg (94 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
Frequently Bought Together