ABS® Upright Refrigerators, Certified to the NSF/ANSI 456 Standard for Vaccine Storage, Horizon Scientific
Fournisseur: Horizon Scientific
Certificats
À propos de cet article
These laboratory and pharmaceutical refrigerators offer premium protection for vaccine storage.
- Microprocessor controller with LED display
- Integrated audible and visual alarms
- Hermetic, high performance compressor
- White powder coated steel construction
- Natural hydrocarbon refrigerants
- NSF/ANSI 456 Vaccine Storage Certification
- ENERGY STAR certified*
These premier refrigerators are designed in accordance with the NSF/ANSI 456 Standard for Vaccine Storage. Units protect pharmaceuticals at optimal temperatures, preventing waste and allowing for peak delivery. The refrigerators utilize microprocessor controllers and feature temperature alarms, remote alarm contacts, and probe access ports with included probes. Units utilize HFC-free refrigerant for environmental health and energy efficiency.
Certifications: Compliant with the thermal performance requirements as defined in the NSF/ANSI 456 Standard for Vaccine Storage for all testing protocols. UL, C-UL, ETL, C-ETL listed (either single or dual agency listings) and certified to UL471 standard, hydrocarbon refrigerant safety.
*Select models ENERGY STAR certified: 76637-278, 76637-282, 76637-306, 76637-284, 76637-308, 76637-286, 76637-310, 76637-290, 76637-314, 76637-292, 76637-316, 76637-294, and 76637-318
*Select models ENERGY STAR certified: 76637-278, 76637-282, 76637-306, 76637-284, 76637-308, 76637-286, 76637-310, 76637-290, 76637-314, 76637-292, 76637-316, 76637-294, and 76637-318
Delivery information: Supplied with screw cap and pouring ring.
Packaging: The bottles are packaged in shrinked-wrapped modules.
Spécifications
- Catalog No:
- 76637-310
- 76637-286
- 76637-278
- 76637-280
- 76637-288
- 76637-302
- 76637-282
- 76637-290
- 76637-304
- 76637-312
- 76637-284
- 76637-292
- 76637-306
- 76637-314
- 76637-294
- 76637-316
- 76637-318
- 76637-308
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Cat. No.:
- 76637-310
- 76637-286
- 76637-278
- 76637-280
- 76637-288
- 76637-302
- 76637-282
- 76637-290
- 76637-304
- 76637-312
- 76637-284
- 76637-292
- 76637-306
- 76637-314
- 76637-294
- 76637-316
- 76637-318
- 76637-308
- Temperature:
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- 1...10 °C
- Defrost:
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Cycle defrost
- Shelves/drawers:
- Fourteen shelves (twelve adjustable/two fixed)
- Fourteen shelves (twelve adjustable/two fixed)
- Seven shelves (six adjustable/one fixed)
- Six shelves (five adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Seven shelves (six adjustable/one fixed)
- Six shelves (five adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Six shelves (five adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Seven shelves (six adjustable/one fixed)
- Five shelves (four adjustable/one fixed)
- Six shelves (five adjustable/one fixed)
- Four shelves (three adjustable/one fixed)
- Ten shelves (eight adjustable/two fixed)
- Five shelves (four adjustable/one fixed)
- Ten shelves (eight adjustable/two fixed)
- Seven shelves (six adjustable/one fixed)
- Door type:
- 2 solid doors
- 2 glass doors
- 1 glass door, right hinged
- 1 glass door
- 1 glass door
- 1 solid door
- 1 glass door
- 1 glass door
- 1 solid door
- 1 solid door
- 1 glass door
- 1 glass door
- 1 solid door
- 1 solid door
- 2 glass doors
- 1 solid door
- 2 solid doors
- 1 solid door
- Weight:
- 205 kg (452 lbs.)
- 247 kg (544 lbs.)
- 86 kg (189 lbs.)
- 120 kg (264 lbs.)
- 120 kg (264 lbs.)
- 86 kg (189 lbs.)
- 131 kg (289 lbs.)
- 131 kg (289 lbs.)
- 116 kg (256 lbs.)
- 116 kg (256 lbs.)
- 164 kg (361 lbs.)
- 164 kg (361 lbs.)
- 128 kg (283 lbs.)
- 128 kg (283 lbs.)
- 247 kg (544 lbs.)
- 127 kg (281 lbs.)
- 205 kg (452 lbs.)
- 127 kg (281 lbs.)
- Environmental certification:
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants
- Natural hydrocarbon refrigerants