Waste Containment Buckets, Perfex
Fournisseur: Perfex
Certificats
A propos de cet article
Temperature-resistant polypropylene bucket with 9 Liter capacity for waste containment.
- Embossed graduations for accurate measure
- Non-porous, super-smooth surface prevents microbial growth
- Compatible with gamma, ethylene oxide, and autoclave sterilization – up to 250˚F for 30 minutes
For use with TruCLEAN 36 Liter bucket, positioned beneath wringer or sieve to capture and isolate contaminants from cleaning and disinfecting agents.
Customers Who Bought This Also Bought