Polycarbin® Racked LTS® Compatible Pipette Tips
Fournisseur: POLYCARBIN, INC.
Certificats
A propos de cet article
Polycarbin’s pipette tip rack is made of 100% circular economy, post-consumer recycled content, combining high performance with a significantly reduced carbon footprint (tips do not contain recycled content).
- Rainin LTS® compatible
- Carries a smaller carbon footprint
- DNase, RNase and pyrogen free
- Sterile
Polycarbin's line of low-carbon lab products are designed for the sustainability-minded scientist. When designing our line of pipette tips, we focused on the the tip box, or 'rack,' which is the most dense, carbon-intensive piece of plastic. All tip boxes, of any brand, can be remanufactured and transformed into the next line of lab products using Polycarbin's alpha carbin program, a mailback service (sold separately).
Certifications: Certified sterile and DNase, RNase and pyrogen free. Manufactured in an ISO 9001 compliant facility.
Packaging: Pipette tips are sold by the case. Each case contains 5 packs of 10 racks (50 racks total, 4800 tips).
Spécifications
- Catalog No:
- 76635-488
- 76635-482
- 76635-498
- 76635-490
- 76635-484
- 76635-508
- Environmentally Preferable:
- Sustainable Packaging, Waste Reducing, Product Transparency and Disclosure
- Sustainable Materials, Safer Human and Environmental Health, Low Manufacturing Impact, Energy Efficient, Waste Reducing, Product Transparency and Disclosure, Water Efficient
- Sustainable Packaging, Waste Reducing, Product Transparency and Disclosure
- Sustainable Packaging, Waste Reducing, Product Transparency and Disclosure
- Sustainable Packaging, Waste Reducing, Product Transparency and Disclosure
- Sustainable Packaging, Waste Reducing, Product Transparency and Disclosure
- Environmental certification:
- This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.
- We base our estimations on a 2018 LCA from the Association of Plastic recyclers. This LCA measures the emissions reductions of recycling 1lb of polypropylene 1 time which confers a 71% reduction in emissions when compared to virgin material.
- This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.
- This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.
- This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.
- This environmental impact of this product was evaulated by a third party, panel reviewed LCA to determine recycled content, carbon dioxide emission eq. reduction, water conservation, and crude oil conservation.