VWR® Sterile Nitrile Cleanroom Class 10 Gloves
Fournisseur: VWR International
Certificats
A propos de cet article
Powder-free gloves designed to provide comfort and tactile sensitivity.
- Hand-specific, beaded cuff
- Silicone-free
- Low particle count
- Specialized manufacturing process
VWR® Sterile nitrile cleanroom class 10 gloves are entirely latex-free to prevent the risk of allergic reactions. These gloves are supplied sterile.
Packaging: Each pair is double packaged in a wallet within a pouch, 20 pouches per bag, 10 bags per case (200 pairs).
Spécifications
- Catalog No:
- 76533-230
- 76533-228
- 76533-226
- 76533-218
- 76533-224
- 76533-222
- 76533-220
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff style:
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Beaded
- Sterility:
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile
- Sterile