Metal Sieves
# de catalogue 470346-962
Fournisseur: United Scientific Supplies
Spécifications
- Description:Metal sieves, set of 2 (4000 and 500 µm)
- Cat. no.:470346-962
Spécifications
A propos de cet article
Quickly sort particles of 4000 and 500 μm inside.
- Stainless steel
- Lid to contain dust
- Aligned with GLOBE protocols
These stackable stainless steel mesh sieves are ideal for separating soil or sediment particles of multiple sizes. Available in six convenient sizes with a lid and catch pan. All sieves are 20 cm in diameter and 4 cm deep.
Customers Who Bought This Also Bought