Piezoelectric Buzzer
# de catalogue 470347-894
Fournisseur: United Scientific Supplies
Spécifications
- Description:Piezoelectric buzzer, 1 - 3 V
- Cat. no.:470347-894
Spécifications
A propos de cet article
Enhance your circuits with sound.
- Requires low voltage 1 - 3 V
- Long leads
This piezoelectric buzzer is an electronic sounder with integration structure. Using a 1 – 3 V DC power supply this can be easily integrated into any electrical circuit. The wire leads can be easily connected to other electrical components.
Customers Who Bought This Also Bought