ContactUsLinkComponent
Piezoelectric Buzzer
Piezoelectric Buzzer
# de catalogue 470347-894
PIEZOELECTRIC BUZZER 1-3V
Piezoelectric Buzzer
# de catalogue 470347-894
Spécifications
  • Description:
    Piezoelectric buzzer, 1 - 3 V
  • Cat. no.:
    470347-894
Spécifications
A propos de cet article
Enhance your circuits with sound.
  • Requires low voltage 1 - 3 V
  • Long leads
This piezoelectric buzzer is an electronic sounder with integration structure. Using a 1 – 3 V DC power supply this can be easily integrated into any electrical circuit. The wire leads can be easily connected to other electrical components.
Customers Who Bought This Also Bought