Zener Diode
# de catalogue 470347-906
Fournisseur: United Scientific Supplies
Spécifications
- Description:Zener diode
- Cat. no.:470347-906
Spécifications
A propos de cet article
Control the current direction in circuits.
- 2 pin connectors
- Fits all standard bred boards
Zener diodes are a unique electrical component that allows the current to flow in a “reverse direction”. A diode only allows a current to flow in one direction and prevents an electrical charge being reversed. A Zener diode does allow the current flow to be reversed, but only once the voltage equals the specific voltage rating.
Customers Who Bought This Also Bought