REUZ™ Stainless Steel Funnels
Fournisseur: United Scientific Supplies
Certificats
A propos de cet article
Stainless steel funnel within air channel for smooth flow.
- 304 stainless steel
- Air channel for smooth flow
- Easy to clean
- Impact resistant
- Available in diameters from 50 to 300 mm
Manufactured with 304 stainless steel, REUZ™ ('re-use') stainless steel funnels include an air channel engineered for smooth, easy flow. Useful in laboratory or factory environments, the versatile funnels can be used with most liquids and powders. Carefully constructed, the funnels are free of sharp edges and easy to clean. Include a convenient stainless steel handle.
REUZ™ products promote sustainability in all laboratory environments.
Ordering information: Sold individually.
Customers Who Bought This Also Bought