Sol-M® Hypodermic Needles
Fournisseur: SOL-MILLENNIUM MEDICAL
New Product
Certificats
A propos de cet article
The Sol-M hypodermic needle product line features a variety of needle length and gauge sizes.
- Triple-bevel lubricated stainless steel bevel that minimizes patient discomfort
- Color-coded hubs for easy gauge identification
- Compatible with all standard luer-lock and slip tip syringes
Sol-Millennium® offers a wide range of hypodermic needles with an emphasis on ease of use and patient comfort.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.