Sol-Vet® Standard Insulin Syringes (Veterinary Care), Sol M
Fournisseur: SOL-MILLENNIUM MEDICAL
Certificats
A propos de cet article
The Sol-Vet Insulin Syringe provides accurate and comfortable insulin administration.
- Bold and clear graduated insulin unit markings for accurate dosing
- Low dead space - improved accuracy and less medication waste when compared to conventional syringes
- Half unit markings
Products for veterinary use only.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.