Sol-M® Standard Pen Needles
Fournisseur: SOL-MILLENNIUM MEDICAL
Certificats
A propos de cet article
Sol-M Pen Needles provide accurate and reliable insulin delivery.
- Double protective cap
- Tri-bevel anti-coring needle to minimize discomfort
- Thin-wall needle for optimal medication flow (31 - 33 G)
- Compatible with most injector pens
They are engineered for comfortable medication delivery with silicone oil lubrication, sharp triple bevel, and thin wall technology.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.