Sol-M® Standard Syringes with Needle
Fournisseur: SOL-MILLENNIUM MEDICAL
Certificats
A propos de cet article
Standard needle and syringe combination.
- High-transparency barrel to easily visualize contents
- Silicone lubrication ensures smooth plunger movement
- Plunger backstop that prevents accidental withdrawal of plunger from the barrel
- Large finger grips allow for easy, controlled injections
The Sol-M syringe with needle features low dead space technology that helps reduce medication waste and improve dosing accuracy.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.