A propos de cet article
The Sol-M Blunt Fill Needle reduces the risk of needlestick injuries to healthcare workers when drawing up and preparing medication by removing the need for sharps.
- Eliminates the unnecessary use of sharps
- Color-coded hubs for easy identification
- Needle hub is compatible with all Luer Lock and Luer Slip Tip syringes
- Available with 5-micron filter to prevent the drawing of glass particles when using ampules
This can be used with the ‘Double Needle Technique’ when drawing medication and can also be used for IV push administration in needleless IV systems.