Sol-M® Multi-Sample Needles
Fournisseur: SOL-MILLENNIUM MEDICAL
Certificats
A propos de cet article
Sol-M Multi-Sample Blood Collection Needles allows for the collection of blood into multiple vacuum collection tubes during a single venipuncture.
- Black dot on multi-sample needle hub ensures bevel up orientation
- Low-angle bevel design allows easy penetration
- Color-coded caps and hubs allow easy identification of needle gauge size
- Premium rubber sheathed needle point for maximum multi-puncture sealing capability
Compatible with all standard holders.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.