A propos de cet article
Gowns are available in Levels 1 through 3 to protect against fluid exposures. Non-surgical isolation gowns are designated as Class I medical devices by the FDA and protect wearers from the transfer of microorganisms and bodily fluids. These gowns are not intended for use in surgeries or invasive procedures.
- Neck tie closures (Level 1 and Level 2 garments)
- Hook and loop neck closures (Level 3 garments)
- Elastic cuffs
- Belt ties
Level 1 AAMI isolation gowns are breathable with ties that allow an adjustable fit. AAMI Level 1 isolation gowns are made from a breathable SMS material with familiar tie design and elastic cuffs.
Level 2 AAMI isolation gowns are made from a premium SMS material that is both breathable and fluid-resistant. Over-the-head construction makes donning quick and convenient. Neck seam is designed for easy tear-away that makes doffing clean and safe for the wearer.
Level 3 AAMI isolation gowns are constructed from poly-coated SPP, which is highly fluid-resistant. They feature a hook and loop neck closure for easy donning and doffing with durable binding. Premium knit cuffs and wide belt ties at the waist.
Spécifications
- Catalog No:
- 76582-784
- Pockets:
- Without pockets
- Gender:
- Unisex
- Garment application:
- Food, Industrial, Cleanroom, Oils, fats and fuels, Laboratory