VWR® Anti-Slip Pads for Caster Wheels
# de catalogue 76407-214
Fournisseur: VWR International
Spécifications
- Description:Anti-slip pads for caster wheels, anti-slip rubber
- Cat. no.:76407-214
Spécifications
A propos de cet article
These anti-slip pads help stabilize and immobilize refrigerators and freezers with casters.
Customers Who Bought This Also Bought