ContactUsLinkComponent
EnzyFluo™ NADP+/NADPH Assay Kit, BioAssay Systems
EnzyFluo™ NADP+/NADPH Assay Kit, BioAssay Systems
# de catalogue 76545-526
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
KIT ASSAY NADP/NADPH
EnzyFluo™ NADP+/NADPH Assay Kit, BioAssay Systems
# de catalogue 76545-526
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    100 tests
Spécifications
A propos de cet article
Direct Assays: NADP+/NADPH concentrations and ratios in cell or tissue extracts.
  • Sensitive and accurate: Detection limit 0.1 µM, linearity up to 10 µM NADP+/NADPH in 96-well plate assay
  • Convenient: The procedure involves adding a single working reagent, and reading the optical density at time zero and 30 min at room temperature
  • High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: Colorimetric Determination at 565 nm
  • Samples: Cells, Tissue
  • Species: human, mouse, rat and more
Pyridine nucleotides play an important role in metabolism and, thus, there is continual interest in monitoring their concentration levels. Quantitative determination of NADP+/NADPH has applications in research pertaining to energy transformation and redox state of cells or tissue.
Simple, direct and automation-ready procedures for measuring NADP+/NADPH concentration are very desirable. BioAssay Systems' EnzyChromTM NADP+ /NADPH assay kit is based on a glucose dehydrogenase cycling reaction, in which the formed NADPH reduces a formazan (MTT) reagent. The intensity of the reduced product color, measured at 565 nm, is proportionate to the NADP+/NADPH concentration in the sample. This assay is highly specific for NADP+/NADPH and is not interfered by NAD+/NADH. Our assay is a convenient method to measure NADP, NADPH and their ratio.
Customers Who Bought This Also Bought