ContactUsLinkComponent
QuantiChrom™ HRP Detection Reagent, BioAssay Systems
QuantiChrom™ HRP Detection Reagent, BioAssay Systems
# de catalogue 76545-500
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
KIT HRP DETECTION REAGENT
QuantiChrom™ HRP Detection Reagent, BioAssay Systems
# de catalogue 76545-500
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    200 tests
Spécifications
A propos de cet article
Direct assays of peroxidases, ELISA, immunochromogenic chemistry, western blot.
  • Method: OD 450 nm
  • Samples: Peroxidases and HRP in ELISA
  • Species: All
  • Detection limit: 76 nU/well, or 7.7 pg/well
HORSERADISH PEROXIDASE (HRP) is an enzyme commonly used when conjugated to analytes of interest. When combined with hydrogen peroxide and a dye, HRP can be used to colorimetrically quantify the analyte.
Customers Who Bought This Also Bought