ContactUsLinkComponent
QuantiChrom™ Salicylate Assay Kit, BioAssay Systems
QuantiChrom™ Salicylate Assay Kit, BioAssay Systems
# de catalogue 76545-496
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
KIT ASSAY SALICYLATE
QuantiChrom™ Salicylate Assay Kit, BioAssay Systems
# de catalogue 76545-496
Fournisseur:  BIOASSAY SYSTEMS
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Size:
    100 tests
  • Cat. no.:
    76545-496
Spécifications
A propos de cet article
Direct Assays: Salicylate in biological samples and beauty products, mouthwash, etc.
  • Fast and sensitive: Linear detection range 0.8 to 20 mM (10.9 to 274.2 mg/dL) salicylate with 20 µl sample (96-well)
  • Convenient: The procedure involves adding a single working reagent
  • High-throughput: 'Add-mix-read' type assay
  • Can be readily automated as a high-throughput 96-well or 384-well plate assay for thousands of samples per day
  • Method: OD 560 nm
  • Samples: Serum, plasma, urine, food, beverage, agriculture, drugs, beauty, mouthwash samples, etc
SALICYLATE is a salt or ester of salicylic acid, and can be found naturally in some plants. It is also a metabolic byproduct of aspirin (acetylsalicylic acid) and salicylate concentrations are often tested in blood or urine in cases of suspected overdose. Salicylic acid is commonly used in skincare products as an exfoliating ingredient, and in other consumer products as a preservative. BioAssay Systems salicylate detection kit provides a convenient and reliable means to measure salicylate.
In the assay, salicylate complexes with ferric chloride to create a colored compound that can be measured at 560 nm. This assay can be used with a variety of samples and is simple, sensitive, and adaptable to high-throughput screening.
Customers Who Bought This Also Bought