LABOPORT™ UN840 Vacuum Diaphragm Pumps, KNF
Fournisseur: KNF Neuberger Inc.
Certificats
A propos de cet article
Chemically resistant, oil-free diaphragm vacuum pump used for a wide range of laboratory applications. Especially suited for handling aggressive vapors, it features PTFE heads, PTFE-coated diaphragms and FFPM valves.
- Handle corrosive and aggressive vapors without damage
- 100% oil-free transfer
- Highly compatible with vapours and condensation
- Chemically resistant
- Suitable for highly corrosive gases and vapours
- High level of gas tightness
Pure transferring and evacuation of gases.
Spécifications
- Catalog No:
- 14224-138
- 26678-016
- 26678-024
- Model:
- N840.1.2FTP
- N840.3FTP
- N840.1.2FTP
- Vacuum:
- 68 torr
- 6 torr
- 68 torr
- Protection class:
- IP 44 enclosure rating
- IP 44 enclosure rating
- IP 44 enclosure rating
- W×D×H:
- 341×160×226 mm
- 330.2×171.5×222.3 mm
- 341×160×226 mm