N920 Diapragm Vacuum Pump, KNF
# de catalogue 97014-794
Fournisseur: KNF Neuberger Inc.
Spécifications
- Protection class:IP 20 enclosure rating
- Vacuum:1.5 torr
- W×D×H:324×158×226 mm
- Description:Diapragm vacuum pump, 920KT.29.18
- Flow rate:20 L/min
- Cat. no.:97014-794
- Electrical:115 V, 60 Hz
- Supplier No.:N920KT.29.18
Spécifications
A propos de cet article
Diaphragm stabilization system allows high suction speed even in the low-vacuum range.
- Features an integrated potentiometer to ensure desired pumping rate selecting appropriate speed
- 100% pure oil-free transfer, evacuation and compression without contaminating the medium
- With PPS pump head, PTFE coated diaphragm, FFPM valves
- Three stage pump head improve leak-tightness
- Gas tight, leakage rate: 6×103 mbar
- Maintenance-free, environmentally friendly pump with adjustable flow rate and thermal switch
- Universal power requirement
- For slightly aggressive or corrosive gases and vapors
Optional analog signal input available for external control.
Customers Who Bought This Also Bought